Learn More
IBA Lifesciences CD45RA Isolation Kit for FABian™
For automated positive cell separation. The Isolation Kit for FABian™ provides label-free target cells without the use of high affinity antibodies and magnetic beads
Brand:  IBA Lifesciences 6-6007-010
Additional Details : Weight : 1.05000kg
 View more versions of this product
Product Code. 16235350
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • The kit is used for the selection of CD45RA+ T cells from whole blood or buffy coat in conjunction with the FABian&trade system.
  • It includes CD45RA Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian&trade run to isolate cells from 9 ml whole blood.
  • Specifications
    Specifications
    Isolation of label-free cells
    2°C to 8°C
    9 mL Whole Blood
    Non-magnetic Cell Isolation Kit
    Human Whole Blood or Buffy Coat
    10 Kits, Human