Learn More
CD45 Mouse, anti-Human, FITC, Clone: HI30, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60018FI.1
Encompass_Preferred
 View more versions of this product
Product Code. 17188491
  • £122.85 / 25 tests
Description
  • The HI30 antibody reacts with all isoforms of CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets, and plasma cells; expression of CD45 is lost during differentiation of these cell types
  • CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
  • The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052) and EasySep™ Human CD45 Depletion Kit II (Catalog #17898; partial blocking may be observed)
Specifications
Specifications
CD45
Monoclonal
FITC
25 Tests
Chimpanzee, Human
Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Leukocytes, Lymphocytes, Myeloid Cells
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
HI30
Mouse
Primary
IgG1 κ