Learn More
CD45 Mouse, anti-Human, FITC, Clone: HI30, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 100-0466
This item is not returnable.
View return policy
Description
- The HI30 antibody reacts with all isoforms of CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets, and plasma cells; expression of CD45 is lost during differentiation of these cell types
- CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
- The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052) and EasySep™ Human CD45 Depletion Kit II (Catalog #17898; partial blocking may be observed)
Specifications
| CD45 | |
| Monoclonal | |
| FITC | |
| 500 Tests | |
| Chimpanzee, Human | |
| Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Leukocytes, Lymphocytes, Myeloid Cells |
| CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| HI30 | |
| Mouse | |
| Primary | |
| IgG1 κ |