missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.1 Mouse, anti-Mouse, Biotin, Clone: A20, STEMCELL Technologies
Mouse Monoclonal Antibody
£98.55 - £205.20
Specifications
| Antigen | CD45.1 |
|---|---|
| Clone | A20 |
| Applications | Immunohistochemistry, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17149141
|
Stemcell Technologies
60117BT.1 |
50 μg |
£98.55
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17139141
|
Stemcell Technologies
60117BT |
500 μg |
£205.20
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The A20 monoclonal antibody reacts with mouse alloantigen CD45.1 (Ly5.1) expressed by all hematopoietic cells except mature erythrocytes and platelets in mouse strains RIII, SJL/J, STS/A, and DA
- Clone A20 does not react with leukocytes that express CD45.2 alloantigen
- CD45.1 is an allelic form of CD45 (leukocyte common antigen), a 180 - 240 kDa type I transmembrane glycoprotein
- CD45 is a member of the protein tyrosine phosphatase (PTP) family and functions in a number of immunoregulatory processes, including cell activation, growth, lymphocyte development, differentiation, and oncogenic transformation
- The isoforms of CD45 correlate to specific cell types, as well as specific activation and maturation states of immune cells
- The A20 monoclonal antibody has been used extensively in adoptive cell transfer and bone marrow transplantation studies in mice, allowing for the differentiation of CD45.2- and CD45.1- expressing cells
Specifications
| CD45.1 | |
| Immunohistochemistry, Flow Cytometry, Immunofluorescence | |
| Biotin | |
| Mouse | |
| Primary |
| A20 | |
| Monoclonal | |
| Mouse | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title