Learn More
CD45.1 Mouse, anti-Mouse, Biotin, Clone: A20, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60117BT.1
This item is not returnable.
View return policy
Description
- The A20 monoclonal antibody reacts with mouse alloantigen CD45.1 (Ly5.1) expressed by all hematopoietic cells except mature erythrocytes and platelets in mouse strains RIII, SJL/J, STS/A, and DA
- Clone A20 does not react with leukocytes that express CD45.2 alloantigen
- CD45.1 is an allelic form of CD45 (leukocyte common antigen), a 180 - 240 kDa type I transmembrane glycoprotein
- CD45 is a member of the protein tyrosine phosphatase (PTP) family and functions in a number of immunoregulatory processes, including cell activation, growth, lymphocyte development, differentiation, and oncogenic transformation
- The isoforms of CD45 correlate to specific cell types, as well as specific activation and maturation states of immune cells
- The A20 monoclonal antibody has been used extensively in adoptive cell transfer and bone marrow transplantation studies in mice, allowing for the differentiation of CD45.2- and CD45.1- expressing cells
Specifications
| CD45.1 | |
| Monoclonal | |
| Biotin | |
| 50 μg | |
| Mouse | |
| Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Lymphocytes, Myeloid Cells |
| Immunohistochemistry, Flow Cytometry, Immunofluorescence | |
| A20 | |
| Mouse | |
| Primary | |
| IgG2a κ |