missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43 Mouse, anti-Human, APC, Clone: CD43-10G7, STEMCELL Technologies
Mouse Monoclonal Antibody
£160.65 - £414.45
Specifications
| Antigen | CD43 |
|---|---|
| Clone | CD43-10G7 |
| Applications | Immunoprecipitation, Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17138941
|
Stemcell Technologies
60085AZ.1 |
25 Tests |
£160.65
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17128941
|
Stemcell Technologies
60085AZ |
100 Tests |
£414.45
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The CD43-10G7 antibody reacts with human CD43, a type I transmembrane sialoglycoprotein expressed on the surface of most human hematopoietic cells, including thymocytes, peripheral T cells, NK cells, monocytes, granulocytes, some B cells (activated B cells and an early B cell subset in bone marrow), plasma cells, and hematopoietic progenitor cells
- Low expression levels are observed on platelets
- CD43 exhibits different glycoforms in the range of ∼95 - 135 kDa, depending on the cell type
- It binds ligands such as CD54 and CD169 (Siglec-1) and plays regulatory roles in cell adhesion and activation, apoptosis and tumor progression, and is used a marker for discriminating normal from neoplastic T cells and B cells within tissues
- Binding of the CD43-10G7 antibody to CD43 on neutrophils reportedly induces homotypic cell adhesion
Specifications
| CD43 | |
| Immunoprecipitation, Flow Cytometry, Functional Assay | |
| APC | |
| Human | |
| Primary |
| CD43-10G7 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title