missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD41 Mouse, anti-Human, APC, Clone: HIP8, STEMCELL Technologies
Mouse Monoclonal Antibody
£252.45 - £533.25
Specifications
| Antigen | CD41 |
|---|---|
| Clone | HIP8 |
| Applications | CyTOF, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17149131
|
Stemcell Technologies
60114AZ.1 |
25 Tests |
£252.45
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17139131
|
Stemcell Technologies
60114AZ |
100 Tests |
£533.25
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The HIP8 antibody reacts with the ∼125 kDa GPIIbα subunit of human CD41, a heterodimeric integrin expressed primarily on platelets, megakaryocytes, monocytes, and acute megakaryoblastic leukemia cells
- The GPIIbα subunit is linked to a smaller ∼25 kDa subunit (GPIIbβ) by a single disulfide bond
- Three isoforms of CD41 that are generated by alternative splicing have been identified, one of which is expressed only by cancerous cells
- CD41 associates non-covalently with CD61 (integrin b3) in a calcium-dependent manner to form the CD41/CD61 (or GPIIb/IIIa) complex, which has been identified as a receptor for several ligands, including fibrinogen, fibronectin, vitronectin, and thrombin
- Binding to the CD41/CD61 complex involves an RGD tripeptidyl sequence in the ligand
- The HIP8 antibody reportedly blocks platelet aggregation and inhibits the activation of platelets by several compounds
Specifications
| CD41 | |
| CyTOF, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay | |
| APC | |
| Baboon, Cynomolgus, Pig, Human | |
| Primary |
| HIP8 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title