missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD41/CD61 Rabbit anti-Human, Clone: BAG 75E5, Novus Biologicals™
Rabbit Monoclonal Antibody
£200.00 - £463.00
Specifications
| Antigen | CD41/CD61 |
|---|---|
| Clone | BAG 75E5 |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18336712
|
Bio-Techne
NBP3-12067-0.025MG |
0.025 mg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18315393
|
Novus Biologicals
NBP3-12067-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD41/CD61 Monoclonal antibody specifically detects CD41/CD61 in Human samples. It is validated for Immunohistochemistry, ImmunofluorescenceSpecifications
| CD41/CD61 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
| This antibody was raised by immunising mice with human CD41/CD61. | |
| Primary |
| BAG 75E5 | |
| Monoclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 3674 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title