missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD41/CD61 Rabbit anti-Human, Clone: BAG 75E5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12067-0.2MG
This item is not returnable.
View return policy
Description
CD41/CD61 Monoclonal antibody specifically detects CD41/CD61 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence
Specifications
| CD41/CD61 | |
| Monoclonal | |
| 1 mg/ml | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
| This antibody was raised by immunising mice with human CD41/CD61. | |
| 0.2 mg | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
| 3674 | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
| IgG κ |
| Immunohistochemistry, Immunofluorescence | |
| BAG 75E5 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction