missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse, anti-Human, PE, Clone: SK3, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60122PE.1
This item is not returnable.
View return policy
Description
- The SK3 antibody reacts with CD4, an ∼59 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains (D1 - D4)
- The epitope for SK3 has been localized to the D3 domain of the protein, which has a structure resembling an Ig variable domain
- CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T-helper cells), and at lower levels by peripheral blood monocytes and macrophages
- CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the affinity of the association between the TCR and MHC II-antigen complex
- CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
- Binding of the SK3 antibody blocks HIV binding and mixed lymphocyte reaction (MLR)
Specifications
| CD4 | |
| Monoclonal | |
| PE | |
| 25 Tests | |
| Human | |
| T Cells, T Cells, CD4+, T Cells, Regulatory |
| Flow Cytometry | |
| SK3 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction