missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human, BUV805, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
£231.60 - £488.81
Specifications
| Antigen | CD4 |
|---|---|
| Clone | SK3 (also known as Leu3a) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 805 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17183664
|
BD Biosciences
612888 |
25 Tests |
£235.00 £231.60 / 7.20g Save £3.40 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16659485
|
BD Biosciences
612887 |
100 Tests |
£494.00 £488.81 / Each Save £5.19 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.Specifications
| CD4 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 805 | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
| SK3 (also known as Leu3a) | |
| Monoclonal | |
| Mouse | |
| Human | |
| Primary | |
| CD4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title