missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human, BUV805, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612888
This item is not returnable.
View return policy
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD4 | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| 25 Tests | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| SK3 (also known as Leu3a) | |
| Mouse | |
| RUO | |
| CD4 | |
| Refrigerate (2 to 8°C) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction