missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD4+ CD45RA+ naive T cells Fab Streptamer™ Isolation Kit MB

Product Code. 15613988 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15613988

Brand: IBA Lifesciences 68000221

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CD4+ CD45RA+ naive T cells Fab Streptamer Isolation Kit MB is for reversible cell isolation delivering label-free target cells.

  • Strep-Tactin™ Magnetic Microbeads for Fab Streptamers™; 3 vials for 5 x 108 cells each
  • CD4 Fab-Strep, human for isolation of T-cells; 1 vial for 1 x 109 cells
  • CD45RA Fab-Strep, human for isolation of T-cells; 1 vial for 5 x 108 cells
  • Biotin stock solution for Streptamer™ technology; 3 x 1 mL
  • Buffer IS, Streptamer™ Washing Buffer, 10x concentrated; 2 x 50 mL
  • Isolation of human CD4+ CD45RA+ T cells is performed by serial double positive selection.
  • In a first step PBMCs are labeled with magnetic CD4 Fab Streptamers™.
  • Labeled cells are isolated in a strong magnet (StrepMan, horizontal position) where they migrate toward the tube wall on the side of the magnet.
  • This CD4 positive cell fraction is collected and cells are liberated from all labeling reagents by addition of biotin in a strong magnet (StrepMan, upright position).
  • The magnetic Streptamers™ migrate toward the tube wall and the label-free cells remain in the supernatant.
  • Biotin is removed by washing.The pure and label-free CD4 positive cells are subjected to a second positive selection with magnetic CD45RA Fab Streptamers™ following the same procedure.
  • The resulting cell preparation is highly enriched with CD4+ CD45RA+ T cells with a purity of more than 90%.
  • No columns are needed.

Specifications

Material PBMCs
Quantity 1 Kit
Type Cell Isolation Kit
Sufficient For 1.5 x 109 cells
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.