missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (GK1.5) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 5 publications
£173.00 - £390.00
Tekniske data
| Antigen | CD4 |
|---|---|
| Clone | GK1.5 |
| Dilution | Flow Cytometry 2.5 ug/ 1 million cells. Use reported in scientific literature (PMID 25340039), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay reported in scientific literature (PMID 25340039), In vivo assay reported in scientific literature (PMID 25340039), Flow (Cell Surface) 2.5 ug/ 1 million cells, Block/Neutralize reported in scientific literature (PMID 25340039) |
| Gene Symbols | CD4 |
| Produktkode | Brand | dimQuantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | dimQuantity | Pris | Antal & tilgængelighed | |||||
|
18791614
|
Novus Biologicals
NBP2-25191SS |
£173.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18728333
|
Novus Biologicals
NBP2-25191 |
£390.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Beskrivelse
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), Block/Neutralize.Tekniske data
| CD4 | |
| GK1.5 | |
| Flow Cytometry 2.5 ug/ 1 million cells. Use reported in scientific literature (PMID 25340039), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay reported in scientific literature (PMID 25340039), In vivo assay reported in scientific literature (PMID 25340039), Flow (Cell Surface) 2.5 ug/ 1 million cells, Block/Neutralize reported in scientific literature (PMID 25340039) | |
| CD4 |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel