Learn More
CD4 Antibody (GK1.5) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP2-25191SS
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), Block/Neutralize.
Specifications
| CD4 | |
| GK1.5 | |
| Flow Cytometry 2.5 ug/ 1 million cells. Use reported in scientific literature (PMID 25340039), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay reported in scientific literature (PMID 25340039), In vivo assay reported in scientific literature (PMID 25340039), Flow (Cell Surface) 2.5 ug/ 1 million cells, Block/Neutralize reported in scientific literature (PMID 25340039) | |
| CD4 |