missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39L4/ENTPD5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | CD39L4/ENTPD5 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645455
|
Novus Biologicals
NBP2-99732-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607915
|
Novus Biologicals
NBP2-99732-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD39L4/ENTPD5 Polyclonal antibody specifically detects CD39L4/ENTPD5 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| CD39L4/ENTPD5 | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2 um filtered solution in PBS | |
| 957 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CD39 antigen-like 4, CD39L4NTPDase 5, CD39-like 4, EC 3.6.1, EC 3.6.1.42, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 5, ER-UDPase, GDPase ENTPD5, Guanosine-diphosphatase ENTPD5, MGC163357, MGC163359, NTPDase-5, Nucleoside diphosphatase, PCPH, Pcph proto-oncogene protein, proto-oncogene CPH, UDPase ENTPD5, Uridine-diphosphatase ENTPD5 | |
| Produced in rabbits immunized with purified, recombinant Human CD39L4/ENTPD5 (Accession#: O75356; Met1-His428) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title