missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39L4/ENTPD5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99732-50ul
This item is not returnable.
View return policy
Description
CD39L4/ENTPD5 Polyclonal antibody specifically detects CD39L4/ENTPD5 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| CD39L4/ENTPD5 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| CD39 antigen-like 4, CD39L4NTPDase 5, CD39-like 4, EC 3.6.1, EC 3.6.1.42, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 5, ER-UDPase, GDPase ENTPD5, Guanosine-diphosphatase ENTPD5, MGC163357, MGC163359, NTPDase-5, Nucleoside diphosphatase, PCPH, Pcph proto-oncogene protein, proto-oncogene CPH, UDPase ENTPD5, Uridine-diphosphatase ENTPD5 | |
| Produced in rabbits immunized with purified, recombinant Human CD39L4/ENTPD5 (Accession#: O75356; Met1-His428) | |
| 50 μg | |
| Lipid and Metabolism | |
| 957 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction