missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD37 Antibody (2B8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28869-0.025ml
This item is not returnable.
View return policy
Description
CD37 Monoclonal specifically detects CD37 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CD37 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| CD37 | |
| Purified recombinant fragment of CD37 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 951 | |
| Human | |
| Ascites |
| Flow Cytometry | |
| 2B8 | |
| Flow Cytometry, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin | |
| CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
| Mouse | |
| 31.7 kDa | |
| 0.025 mL | |
| Primary | |
| Specific for CD37. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction