missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD37 Antibody (2B8), Novus Biologicals™
Mouse Monoclonal Antibody
£144.00
Specifications
| Antigen | CD37 |
|---|---|
| Clone | 2B8 |
| Dilution | Flow Cytometry, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD37 Monoclonal specifically detects CD37 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CD37 | |
| Flow Cytometry, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| 951 | |
| Purified recombinant fragment of CD37 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 31.7 kDa |
| 2B8 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
| CD37 | |
| IgG1 | |
| Unpurified | |
| Specific for CD37. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title