missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD366 (TIM3) Monoclonal Antibody (8B.2C12), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£2870.00 - £12950.00
Specifications
| Antigen | CD366 (TIM3) |
|---|---|
| Clone | 8B.2C12 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17181312
|
Invitrogen™
16-5871-38 |
5 mg |
£2870.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17161312
|
Affymetrix eBioscience
16-5871-025 |
25 mg | |||||||
|
17171312
|
Affymetrix eBioscience
16-5871-050 |
50 mg | |||||||
Description
TIM3 (Hepatitis A virus cellular receptor 2, HAVCR2, T-cell immunoglobulin, mucin-dmain containing-3) is a 281 amino acid long, Type-1 Th1- specific cell surface glycoprotein expressed on terminally differentiated CD4+Th1 and CD8+Tc1 cells. TIM3 consists of an IgV-like domain, a mucin-like domain in the extracellular region, and a conserved Tyrosine phosphorylation motif in the cytoplasmic region. TIM3 is involved in macrophage activation and induction of autoimmune diseases. Further, TIM3 down-regulates aggressive Th1-mediated immune responses and facilitates in the development of immune tolerance. Pathological significance of TIM3 has been attributed to Experimental autoimmune encephalomyelitis (EAE), a Th-1 dependent autoimmune disease, and also enhances the severity of experimental autoimmune encephalomyelitis in mice.Specifications
| CD366 (TIM3) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 171285 | |
| Primary | |
| 4°C | |
| Havcr2 |
| 8B.2C12 | |
| Flow Cytometry, Functional Assay | |
| Functional Grade | |
| Rat | |
| Mouse | |
| Q8VIM0 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title