missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD366 (TIM3) Monoclonal Antibody (8B.2C12), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5871-050
This item is not returnable.
View return policy
Description
CD366 (TIM3) Monoclonal antibody specifically detects CD366 (TIM3) in Mouse samples. It is validated for Flow Cytometry, Functional Assay
Specifications
| CD366 (TIM3) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| Q8VIM0 | |
| Rat | |
| 50 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Functional Assay | |
| 8B.2C12 | |
| Functional Grade | |
| Havcr2 | |
| CD366; FLJ14428; Havcr2; HAVcr-2; Hepatitis A virus cellular receptor 2; hepatitis A virus cellular receptor 2 homolog; kidney injury molecule-3; KIM-3; sCD366; soluble CD366; soluble TIM 3; T cell immunoglobulin mucin 3; T cell immunoglobulin mucin-3; T-cell immunoglobulin and mucin domain containing 3; T-cell immunoglobulin and mucin domain-containing protein 3; T-cell immunoglobulin mucin family member 3; T-cell immunoglobulin mucin receptor 3; T-cell membrane protein 3; TIM3; TIM-3; TIMD3; TIMD-3 | |
| Affinity chromatography | |
| RUO | |
| 171285 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction