Learn More
CD34 Mouse anti-Human, PE, Clone: 581, BD
Mouse Monoclonal Antibody
£55.99 - £255.26
Specifications
| Antigen | CD34 |
|---|---|
| Clone | 581 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15850619
|
BD Biosciences
560941 |
25 Tests |
£261.00 £55.99 / Each Save £205.01 78% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10285964
|
BD Biosciences
555822 |
100 Tests |
£259.00 £255.26 / Each Save £3.74 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 581 monoclonal antibody specifically binds to CD34, a sialomucin-like type I transmembrane glycoprotein. This single-chain, 105-120 kDa, heavily O-glycosylated protein is expressed on hematopoietic progenitor cells, vascular endothelium, bone marrow stromal cells and embryonic fibroblasts. The cytoplasmic region of the CD34 antigen is a target for phosphorylation by activated protein kinase C suggesting CD34 may play a role in signal transduction. CD34 may also play a role as an adhesion molecule since it binds to CD62E and CD62L. Clone 581 binds to the class III CD34 epitope, it is resistant to neuraminidase, chymopapain and glycoprotease. The 581 antibody blocks reactivity of another anti-CD34 monoclonal antibody, 8G12.
Specifications
| CD34 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 581 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| gp105-120; My10; Hematopoietic progenitor cell antigen CD34 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.