missing translation for 'onlineSavingsMsg'
Learn More

CD34 Mouse anti-Human, PE, Clone: 581, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560941

 View more versions of this product

Product Code. 15850619

  • ONLINE EXCLUSIVE PRICE
    £261.00
    £55.99 / Each
    Save £205.01
    78% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 581 monoclonal antibody specifically binds to CD34, a sialomucin-like type I transmembrane glycoprotein. This single-chain, 105-120 kDa, heavily O-glycosylated protein is expressed on hematopoietic progenitor cells, vascular endothelium, bone marrow stromal cells and embryonic fibroblasts. The cytoplasmic region of the CD34 antigen is a target for phosphorylation by activated protein kinase C suggesting CD34 may play a role in signal transduction. CD34 may also play a role as an adhesion molecule since it binds to CD62E and CD62L. Clone 581 binds to the class III CD34 epitope, it is resistant to neuraminidase, chymopapain and glycoprotease. The 581 antibody blocks reactivity of another anti-CD34 monoclonal antibody, 8G12.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD34
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Human
IgG1 κ
Flow Cytometry
581
gp105-120; My10; Hematopoietic progenitor cell antigen CD34
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.