Learn More
CD33 Mouse, anti-Human, PE, Clone: P67.6, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60126PE
Encompass_Preferred
 View more versions of this product
Product Code. 17109161
  • £476.55 / 100 tests
Description
  • The P67.6 antibody reacts with CD33 (Siglec-3), a 67 kDa type 1 transmembrane sialoadhesin protein of the immunoglobulin superfamily
  • CD33 is expressed on committed myeloid progenitor cells, monocytes, granulocytes, dendritic cells, mast cells, resident macrophages, subsets of B cells, and activated T and NK cells
  • CD33 is a major marker used to differentiate myeloid from lymphoid or erythroid leukemias
  • In peripheral blood, it is expressed at high levels on monocytes and at lower levels on granulocytes
  • CD33 functions as a sialic acid-dependent cell adhesion molecule and mediates protein-glycan and protein-protein interactions between cells
  • It also has a putative role as an inhibitory receptor during the immune response
Specifications
Specifications
CD33
Monoclonal
PE
100 Tests
Human
Granulocytes and Subsets, Myeloid Cells
Flow Cytometry
P67.6
Mouse
Primary
IgG1 κ