missing translation for 'onlineSavingsMsg'
Learn More

CD33 Mouse, anti-Human, PE, Clone: P67.6, STEMCELL Technologies

Mouse Monoclonal Antibody

£194.40 - £476.55

Specifications

Antigen CD33
Clone P67.6
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17119161
Encompass_Preferred
View Documents
Stemcell Technologies
60126PE.1
25 Tests
£194.40
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17109161
Encompass_Preferred
View Documents
Stemcell Technologies
60126PE
100 Tests
£476.55
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The P67.6 antibody reacts with CD33 (Siglec-3), a 67 kDa type 1 transmembrane sialoadhesin protein of the immunoglobulin superfamily
  • CD33 is expressed on committed myeloid progenitor cells, monocytes, granulocytes, dendritic cells, mast cells, resident macrophages, subsets of B cells, and activated T and NK cells
  • CD33 is a major marker used to differentiate myeloid from lymphoid or erythroid leukemias
  • In peripheral blood, it is expressed at high levels on monocytes and at lower levels on granulocytes
  • CD33 functions as a sialic acid-dependent cell adhesion molecule and mediates protein-glycan and protein-protein interactions between cells
  • It also has a putative role as an inhibitory receptor during the immune response
Specifications

Specifications

CD33
Flow Cytometry
PE
Human
Primary
P67.6
Monoclonal
Mouse
IgG1 κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.