Learn More
CD33 Mouse anti-Human, FITC, Clone: HIM3-4, BD
Mouse Monoclonal Antibody
£44.51 - £156.31
Specifications
| Antigen | CD33 |
|---|---|
| Clone | HIM3-4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15831369
|
BD Biosciences
561818 |
25 Tests |
£51.40 £44.51 / Each Save £6.89 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15828318
|
BD Biosciences
555626 |
100 Tests |
£163.00 £156.31 / Each Save £6.69 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reacts with a 67 kDa type I transmembrane glycoprotein expressed on monocytes, activated T cells, myeloid progenitors as well as mast cells. CD33 is absent on normal platelets, lymphocytes, erythrocytes and hematopoietic stem cells. Reports indicate that this glycoprotein can function as a sialic acid-dependent cell adhesion molecule. This function can be modulated by endogenous sialoglycoconjugates when CD33 is expressed on the membrane. HIM3-4 reacts with human and monkey granulocytes and monocytes.
Specifications
| CD33 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C |
| HIM3-4 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.