Learn More
CD33 Mouse anti-Human, FITC, Clone: HIM3-4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561818
Description
Reacts with a 67 kDa type I transmembrane glycoprotein expressed on monocytes, activated T cells, myeloid progenitors as well as mast cells. CD33 is absent on normal platelets, lymphocytes, erythrocytes and hematopoietic stem cells. Reports indicate that this glycoprotein can function as a sialic acid-dependent cell adhesion molecule. This function can be modulated by endogenous sialoglycoconjugates when CD33 is expressed on the membrane. HIM3-4 reacts with human and monkey granulocytes and monocytes.
Specifications
| CD33 | |
| Monoclonal | |
| FITC | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| HIM3-4 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.