missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32A/C Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CD32A/C |
|---|---|
| Concentration | 900 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066885
|
Proteintech
25323-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18029445
|
Proteintech
25323-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD32A/C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD32A/C. This antibody recognizes human antigen. The CD32A/C antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CD32A/C | |
| Western Blot, ELISA | |
| Rabbit | |
| 9103 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 900 μg/mL | |
| Liquid | |
| Human | |
| CD32, CD32C, FcgammaRIIC, FCGR2B, FCGR2C, FcRIIC, hFcRII C | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title