Learn More
CD32A/C Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25323-1-AP-20UL
This item is not returnable.
View return policy
Description
The CD32A/C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD32A/C. This antibody recognizes human antigen. The CD32A/C antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CD32A/C | |
| 900 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| CD32, CD32C, FcgammaRIIC, FCGR2B, FCGR2C, FcRIIC, hFcRII C | |
| 20 μL | |
| 9103 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |