missing translation for 'onlineSavingsMsg'
Learn More

CD32 Mouse, anti-Human, FITC, Clone: IV.3, STEMCELL Technologies

Mouse Monoclonal Antibody

POA - £207.90

Specifications

Antigen CD32
Clone IV.3
Applications Blocking Assay, Western Blot, Flow Cytometry, Immunocytochemistry, Functional Assay
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17108431
Encompass_Preferred
View Documents
Stemcell Technologies
60012FI.1
25 Tests
£207.90
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17161046
Encompass_Preferred
View Documents
Stemcell Technologies
60012FI
100 Tests This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • The IV.3 antibody reacts with human CD32 (FcγRII), an ∼40 kDa type 1 transmembrane glycoprotein that mediates several functions including phagocytosis, cytotoxicity, immunomodulation and platelet aggregation
  • CD32 is encoded by three genes (A, B, C) and at least 6 isoforms are generated via alternative mRNA splicing, i.e., IIa1, IIa2, IIb1, IIb2, IIb3 and IIc
  • All isoforms are expressed by monocytes/macrophages, placental trophoblasts and endothelial cells
  • In addition, the IIb isoform is expressed by B cells, and the IIa isoform by platelets, granulocytes and, weakly, by B cells
  • Isoform IIc is expressed by NK cells and neutrophils
  • CD32 binds weakly to the Fc region of monomeric IgG but more strongly to IgG aggregates and immune complexes
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051) and EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052)
Specifications

Specifications

CD32
Blocking Assay, Western Blot, Flow Cytometry, Immunocytochemistry, Functional Assay
FITC
Human
Primary
IV.3
Monoclonal
Mouse
IgG2b κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.