missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32 Mouse, anti-Human, FITC, Clone: IV.3, STEMCELL Technologies
Description
- The IV.3 antibody reacts with human CD32 (FcγRII), an ∼40 kDa type 1 transmembrane glycoprotein that mediates several functions including phagocytosis, cytotoxicity, immunomodulation and platelet aggregation
- CD32 is encoded by three genes (A, B, C) and at least 6 isoforms are generated via alternative mRNA splicing, i.e., IIa1, IIa2, IIb1, IIb2, IIb3 and IIc
- All isoforms are expressed by monocytes/macrophages, placental trophoblasts and endothelial cells
- In addition, the IIb isoform is expressed by B cells, and the IIa isoform by platelets, granulocytes and, weakly, by B cells
- Isoform IIc is expressed by NK cells and neutrophils
- CD32 binds weakly to the Fc region of monomeric IgG but more strongly to IgG aggregates and immune complexes
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051) and EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052)
Specifications
Specifications
| Antigen | CD32 |
| Applications | Blocking Assay, Western Blot, Flow Cytometry, Immunocytochemistry, Functional Assay |
| Classification | Monoclonal |
| Clone | IV.3 |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 25 Tests |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG2b κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?