missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD31/PECAM-1 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence procedures.

£110.13 - £401.77

Specifications

Antigen CD31 (Endothelial Cell Marker) Ab-1
Clone JC/70A
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12601137
View Documents
Epredia™
MS-353-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12611137
View Documents
Epredia™
MS-353-S1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
£323.00
£302.76 / 500µL
Save £20.24
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12661127
View Documents
Epredia™
MS-353-S
1 mL Concentrated
ONLINE EXCLUSIVE PRICE
£428.00
£401.77 / 1mL
Save £26.23
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12681127
View Documents
Epredia™
MS-353-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651127
View Documents
Epredia™
MS-353-RQ
12 mL Ready-to-Use
ONLINE EXCLUSIVE PRICE
£349.00
£327.92 / 12mL
Save £21.08
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion.

Host Species: Mouse

Clone: JC/70A

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat and pig. Others not tested.

Epitope: Not determined

Immunogen: Membrane preparation of a spleen from a patient with hairy cell leukemia

Molecular Weight: 100kDa (endothelial cells) and 130kDa (platelets)

Positive Control: Placenta, Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence

Specifications

Specifications

CD31 (Endothelial Cell Marker) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
JC/70A
Monoclonal
Mouse
Oncogenesis
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.