missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD31/PECAM-1 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence procedures.

Brand:  Epredia™ MS-353-S1

 View more versions of this product

Product Code. 12611137

  • ONLINE EXCLUSIVE PRICE
    £323.00
    £302.76 / 500µL
    Save £20.24
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion.

Host Species: Mouse

Clone: JC/70A

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat and pig. Others not tested.

Epitope: Not determined

Immunogen: Membrane preparation of a spleen from a patient with hairy cell leukemia

Molecular Weight: 100kDa (endothelial cells) and 130kDa (platelets)

Positive Control: Placenta, Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD31 (Endothelial Cell Marker) Ab-1
Monoclonal
Unconjugated
Membrane preparation of a spleen from a patient with hairy cell leukemia
IVD
Primary
Concentrated
Immunofluorescence, Immunohistochemistry (Paraffin)
JC/70A
Mouse
500 μL
Oncogenesis
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.