Learn More
IBA Lifesciences CD31 Fab-TACS™ Agarose Column Starter Kit
Label-free isolation of CD31+ cells from whole blood, buffy coat or PBMCs
Brand: IBA Lifesciences 6-3216-002
Additional Details : CAS Number : 26628-22-8 Weight : 0.18600kg
Description
- Fab-TACS™ - Traceless Affinity Cell Selection - is a positive cell isolation approach based on our Strep-tag™ technology
- CD31 Fab-TACS™ Agarose column Starter Kit allows label-free isolation of human CD31+ cells from whole blood, buffy coat, PBMCs or other cell suspensions
- CD31 is also known as platelet endothelial cell adhesion molecule (PECAM-1) and expressed on different cell types including endothelial cells, platelets, monocytes, neutrophils and lymphocytes.
- Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation
- Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix
- Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target cells are efficiently washed away
- In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps
- After elution, the Fab-Streps self-dissociate from the cell surfaces due to their reduced affinity
- The label-free authentic target cells are now ready for further downstream applications
Specifications
Strep-Tactin™ TACS Agarose columns, 1 mL (2 x), CD31 Fab-Strep (human), 10 x Buffer CI, Biotin stock solution, TACS Column Adapter | |
Human |
For Research Use Only.