missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD304 Rat anti-Mouse , BV421, Clone: V46-1954, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 567112
This item is not returnable.
View return policy
Description
CD304 Monoclonal antibody specifically detects CD304 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD304 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| neuropilin-1; Nrp1; A5 protein; NP-1; NPN-1; Npn1; Nrp | |
| Mouse CD304 Recombinant Protein | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| V46-1954 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction