missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD304 Rat anti-Mouse , BV421, Clone: V46-1954, BD Biosciences™
Rat Monoclonal Antibody
£154.08 - £359.52
Specifications
| Antigen | CD304 |
|---|---|
| Clone | V46-1954 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17257834
|
BD Biosciences
567113 |
25 μg |
£161.00 £154.08 / 25µg Save £6.92 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17247834
|
BD Biosciences
567112 |
0.1 mg |
£375.00 £359.52 / 100µg Save £15.48 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD304 Monoclonal antibody specifically detects CD304 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD304 | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse CD304 Recombinant Protein | |
| Primary |
| V46-1954 | |
| Monoclonal | |
| Rat | |
| Mouse | |
| neuropilin-1; Nrp1; A5 protein; NP-1; NPN-1; Npn1; Nrp | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title