missing translation for 'onlineSavingsMsg'
Learn More

CD30 / TNFRSF8 Mouse, Clone: rKi-1/779, enQuireBio™

Mouse Monoclonal Antibody

£181.83 - £423.29

Specifications

Antigen CD30 / TNFRSF8
Clone rKi-1/779
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15961218
View Documents
EnquireBio
943-MSM8-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15971218
View Documents
EnquireBio
943-MSM8-P1
100 μg
£423.29
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD30 / TNFRSF8
200μg/mL
Monoclonal
Mouse
10mM PBS with or without 0.05% BSA and 0.05% azide.
943
Recombinant full-length human TNFRSF8 protein
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
105-120kDa
rKi-1/779
Immunohistochemistry
Unconjugated
RUO
XP_011540745
TNFRSF8
IgG1 κ
Protein A/G Purified
Cell surface
Hodgkin's lymphoma
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.