missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 / TNFRSF8 Mouse, Clone: rKi-1/779, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 943-MSM8-P1
This item is not returnable.
View return policy
Specifications
| CD30 / TNFRSF8 | |
| Cell surface | |
| rKi-1/779 | |
| Unconjugated | |
| XP_011540745 | |
| Mouse | |
| 105-120kDa | |
| Protein A/G Purified | |
| RUO | |
| 943 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| TNFRSF8 | |
| Recombinant full-length human TNFRSF8 protein | |
| Hodgkin's lymphoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction