missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-361-RQ

Code : LD

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 12601187

  • £247.00 / 12mL
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD30, a single chain glycoprotein, is synthesized as a 90 kDa precursor which is processed in the Golgi complex into a membrane-bound phosphorylated mature 105/120kDa glycoprotein. The CD30/Ki-1 antigen is expressed by mononuclear Hodgkin and multinucleated Reed-Sternberg cells in Hodgkin's disease, by the tumor cells of a majority of anaplastic large cell lymphomas, and by a varying proportion of activated T and B cells.

Host Species: Mouse

Clone: Ber-H2

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with baboon and monkey. Others not known.

Epitope: Not determined

Immunogen: Co cells

Molecular Weight: 120 kDa (mature), 95 kDa (precursor)

Positive Control: Hodgkin's lymphoma

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD30 Ab-1
Monoclonal
Unconjugated
Co cells
IVD
Primary
Immunohistochemistry (Paraffin)
Ber-H2
Mouse
12 mL
Oncogenesis
Ready-to-Use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers