missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody.
£106.00 - £391.00
Specifications
Antigen | CD30 Ab-1 |
---|---|
Clone | Ber-H2 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12621187
|
Epredia™
MS-361-S0 |
Concentrated | 100 μL |
£106.00
100µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12631187
|
Epredia™
MS-361-S1 |
Concentrated | 500 μL |
£288.00
500µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12611187
|
Epredia™
MS-361-S |
Concentrated | 1 mL |
£391.00
1mL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12691177
|
Epredia™
MS-361-R7 |
Prediluted | 7 mL |
£209.00
7mL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12601187
|
Epredia™
MS-361-RQ |
Ready-to-Use | 12 mL |
£234.00
12mL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
CD30, a single chain glycoprotein, is synthesized as a 90 kDa precursor which is processed in the Golgi complex into a membrane-bound phosphorylated mature 105/120kDa glycoprotein. The CD30/Ki-1 antigen is expressed by mononuclear Hodgkin and multinucleated Reed-Sternberg cells in Hodgkin's disease, by the tumor cells of a majority of anaplastic large cell lymphomas, and by a varying proportion of activated T and B cells.Host Species: Mouse
Clone: Ber-H2
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with baboon and monkey. Others not known.
Epitope: Not determined
Immunogen: Co cells
Molecular Weight: 120 kDa (mature), 95 kDa (precursor)
Positive Control: Hodgkin's lymphoma
Cellular Localization: Membrane and cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD30 Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Co cells |
Ber-H2 | |
Monoclonal | |
Mouse | |
Oncogenesis | |
Primary |