missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 zeta Rabbit anti-Human, Clone: 1T5R7, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £413.00
Specifications
| Antigen | CD3 zeta |
|---|---|
| Clone | 1T5R7 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18394815
|
Novus Biologicals
NBP3-15349-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18309276
|
Novus Biologicals
NBP3-15349-100UL |
100 μg |
£413.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CD3 zeta | |
| Western Blot, Flow Cytometry, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
| A synthetic peptide corresponding to a sequence within amino acids 65-164 of human CD3 zeta (P20963). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1T5R7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 919 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title