missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 zeta Rabbit anti-Human, Clone: 1T5R7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15349-100UL
This item is not returnable.
View return policy
Description
CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CD3 zeta | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1T5R7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
| A synthetic peptide corresponding to a sequence within amino acids 65-164 of human CD3 zeta (P20963). | |
| 100 μg | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 919 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction