missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse, APC-Cy7, Clone: UCHT1, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB6-APC7-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952248

  • £109.00 / 25 tests
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD3
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD3D
Protein A/G Purified
RUO
915
IgG1 κ
Flow Cytometry
UCHT1
APC-Cyanine7
NP_001035741
Mouse
25 Tests
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers