missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, APC-Cy7, Clone: UCHT1, enQuireBio™
Mouse Monoclonal Antibody
£109.00 - £210.00
Specifications
Antigen | CD3 |
---|---|
Clone | UCHT1 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952248
|
EnquireBio
QAB6-APC7-25TESTS |
25 Tests |
£109.00
25 tests |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942248
|
EnquireBio
QAB6-APC7-100TESTS |
100 Tests |
£210.00
100 tests |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD3 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
915 | |
IgG1 κ | |
Protein A/G Purified |
UCHT1 | |
Flow Cytometry | |
APC-Cyanine7 | |
RUO | |
NP_001035741 | |
CD3D | |
Primary | |
Store at 2-4°C. |