missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, anti-Human, FITC, Clone: SK7, STEMCELL Technologies
Mouse Monoclonal Antibody
£133.65 - £295.65
Specifications
| Antigen | CD3 |
|---|---|
| Clone | SK7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17139161
|
Stemcell Technologies
60127FI.1 |
25 Tests |
£133.65
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17129161
|
Stemcell Technologies
60127FI |
100 Tests |
£295.65
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The SK7 antibody reacts with the ∼20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ∼95% of mature T cells and NKT cells, and variably on thymocytes
- A majority of T cell neoplasms also express CD3
- The CD3 complex, which is assembled from combinations of CD3γ, δ, ε, η and ζ subunits, associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR
- Activation of T cells by the TCR involves the cytoplasmic tails of the CD3 subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily
- Mutations in the CD3 subunits have been associated with various immunodeficiency disorders, including severe combined immunodeficiency (SCID)
Specifications
| CD3 | |
| Flow Cytometry | |
| FITC | |
| Chimpanzee, Human | |
| Primary |
| SK7 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title