Learn More
CD3 Mouse, anti-Human, FITC, Clone: SK7, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60127FI.1
Encompass_Preferred
 View more versions of this product
Product Code. 17139161
  • £133.65 / 25 tests
Description
  • The SK7 antibody reacts with the ∼20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ∼95% of mature T cells and NKT cells, and variably on thymocytes
  • A majority of T cell neoplasms also express CD3
  • The CD3 complex, which is assembled from combinations of CD3γ, δ, ε, η and ζ subunits, associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR
  • Activation of T cells by the TCR involves the cytoplasmic tails of the CD3 subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily
  • Mutations in the CD3 subunits have been associated with various immunodeficiency disorders, including severe combined immunodeficiency (SCID)
Specifications
Specifications
CD3
Monoclonal
FITC
25 Tests
Chimpanzee, Human
T Cells
Flow Cytometry
SK7
Mouse
Primary
IgG1 κ