missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Baboon,Cynomolgus,Rhesus Ma,Human, BB700, Clone: SP34-2, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566518
Additional Details : Weight : 0.00700kg
Description
CD3 Monoclonal antibody specifically detects CD3 in Baboon,Cynomolgus,Rhesus Ma,Human samples. It is validated for Flow Cytometry.Specifications
CD3 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Baboon, Cynomolgus, Rhesus Macaque, Human | |
Purified |
Flow Cytometry | |
SP34-2 | |
Brilliant Blue 700 (BB700) | |
25 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 λ |