missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Baboon,Cynomolgus,Rhesus Ma,Human, BB700, Clone: SP34-2, BD Horizon™
Mouse Monoclonal Antibody
£128.40 - £361.57
Specifications
| Antigen | CD3 |
|---|---|
| Clone | SP34-2 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15875119
|
BD Biosciences
566518 |
25 μg |
£134.00 £128.40 / Each Save £5.60 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15865119
|
BD Biosciences
566517 |
0.1 mg |
£377.00 £361.57 / Each Save £15.43 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 Monoclonal antibody specifically detects CD3 in Baboon,Cynomolgus,Rhesus Ma,Human samples. It is validated for Flow Cytometry.Specifications
| CD3 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 λ | |
| Refrigerate (2 to 8°C) |
| SP34-2 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Baboon, Cynomolgus, Rhesus Macaque, Human | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title