Learn More
Epredia™ Lab Vision™ CD3ε (Early T-Cell Marker) Ab-9, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD3ε (Early T-Cell Marker) Ab-9, Mouse Monoclonal Antibody.
£105.06 - £504.90
Specifications
| Antigen | CD3e |
|---|---|
| Clone | F7.2.38 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12670387
|
Epredia™
MS-1880-S0 |
100 μL | Concentrated | ||||||
|
12680387
|
Epredia™
MS-1880-S1 |
500 μL | Concentrated | ||||||
|
12660387
|
Epredia™
MS-1880-S |
1 mL | Concentrated | ||||||
|
12650387
|
Epredia™
MS-1880-R7 |
7 mL | Prediluted | ||||||
Description
CD3 consists of five different polypeptide chains (designated as gamma, delta, epsilon, zeta, and eta). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells and is present in the majority of T cell neoplasms.Host Species: Mouse
Clone: F7.2.38
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Cytoplasmic region of ε-chain
Immunogen: Purified CD3 ε γ δ/CD3ω
Molecular Weight: ∽20kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| CD3e | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG2a κ |
| F7.2.38 | |
| Monoclonal | |
| Mouse | |
| T-Cell | |
| Purified CD3 epsilon gamma delta/CD3omega | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.