missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD3ε (Early T-Cell Marker) Ab-9, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD3ε (Early T-Cell Marker) Ab-9, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1880-R7

 View more versions of this product

Product Code. 12650387

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD3 consists of five different polypeptide chains (designated as gamma, delta, epsilon, zeta, and eta). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells and is present in the majority of T cell neoplasms.

Host Species: Mouse

Clone: F7.2.38

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Cytoplasmic region of ε-chain

Immunogen: Purified CD3 ε γ δ/CD3ω

Molecular Weight: ∽20kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD3e
Monoclonal
Unconjugated
Purified CD3 epsilon gamma delta/CD3omega
RUO
Primary
Prediluted
Immunohistochemistry (Paraffin)
F7.2.38
Mouse
7 mL
T-Cell
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.